The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028115.3 | NP_082391.1 | 5 - 6 | 606 | 82 |
NM_029839.3 | NP_084115.2 | 5 - 6 | 606 | 82 | |
GenBank mRNA | AK011362.1 | - | 5 - 6 | 601 | 82 |
AK018525.1 | - | 5 - 6 | 579 | 82 | |
AK031674.1 | - | 5 - 6 | 584 | 82 | |
AK164095.1 | - | 5 - 6 | 577 | 82 | |
AK169221.1 | - | 5 - 6 | 572 | 82 | |
BC039287.1 | - | 5 - 6 | 551 | 82 | |
BC054088.1 | - | 5 - 6 | 420 | 82 | |
BC089592.1 | - | 5 - 6 | 573 | 82 |