Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgap1
Entrez Gene ID
241062
Gene Name
post-GPI attachment to proteins 1
Gene Aliases
5033403E17Rik, 9030223K07Rik, A530084K22, D230012E17Rik, oto
Chromosome Location
Chr.1: 54473000 - 54557684 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163314.2 NP_001156786.1 4 - 5 750 111
GenBank mRNA AK041133.1 - 4 - 5 751 111
AK138171.1 - 4 - 5 690 111

View Full Product Details