The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002238.2 | NP_001002238.1 | 3 - 4 | 452 | 101 |
NM_001002240.2 | NP_001002240.1 | 3 - 4 | 425 | 101 | |
NM_001002241.2 | NP_001002241.1 | 2 - 3 | 725 | 101 | |
NM_031387.3 | NP_113564.2 | 3 - 4 | 585 | 101 | |
GenBank mRNA | AB067571.1 | - | 2 - 3 | 406 | 101 |
AB183526.1 | - | 3 - 4 | 452 | 101 | |
AB183527.1 | - | 3 - 4 | 425 | 101 | |
AB183528.1 | - | 2 - 3 | 725 | 101 | |
AB183529.1 | - | 2 - 3 | 289 | 101 | |
AK029806.1 | - | 3 - 4 | 586 | 101 |