Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vil1
Entrez Gene ID
22349
Gene Name
villin 1
Gene Aliases
Vil
Chromosome Location
Chr.1: 74409191 - 74435560 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009509.2 NP_033535.2 5 - 6 513 68
GenBank mRNA AI663456.1 - 5 - 6 505 68
AK027908.1 - 5 - 6 510 68
BC015267.1 - 5 - 6 489 68
BC117875.2 - 5 - 6 478 68
BC145671.1 - 5 - 6 480 68
M98454.1 - 5 - 6 503 68

View Full Product Details