The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011937.2 | NP_036067.2 | 1 - 2 | 82 | 89 |
GenBank mRNA | AF088903.1 | - | 1 - 2 | 89 | |
BY092839.1 | - | 1 - 2 | 57 | 89 | |
CJ048539.1 | - | 1 - 2 | 83 | 89 |