The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145494.2 | NP_663469.1 | 14 - 15 | 1588 | 65 |
GenBank mRNA | AK042289.1 | - | 14 - 15 | 1635 | 65 |
AK050933.1 | - | 14 - 15 | 1635 | 65 | |
AK050980.1 | - | 14 - 15 | 1635 | 65 | |
AK156403.1 | - | 14 - 15 | 1635 | 65 | |
AK171157.1 | - | 14 - 15 | 1635 | 65 | |
BC004709.1 | - | 14 - 15 | 1588 | 65 |