The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110202.1 | NP_001103672.1 | 11 - 12 | 2531 | 80 |
NM_001286387.1 | NP_001273316.1 | 13 - 14 | 2750 | 80 | |
NM_001286388.1 | NP_001273317.1 | 13 - 14 | 2807 | 80 | |
NM_053167.3 | NP_444397.2 | 13 - 14 | 2753 | 80 | |
GenBank mRNA | AK028280.1 | - | 11 - 12 | 2250 | 80 |
AK129109.1 | - | 13 - 14 | 2458 | 80 | |
AK163123.1 | - | 13 - 14 | 2487 | 80 |