The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110202.1 | NP_001103672.1 | 3 - 4 | 1491 | 63 |
NM_001110203.1 | NP_001103673.1 | 3 - 4 | 1491 | 63 | |
NM_001286386.1 | NP_001273315.1 | 3 - 4 | 1491 | 63 | |
NM_001286387.1 | NP_001273316.1 | 3 - 4 | 1491 | 63 | |
NM_001286388.1 | NP_001273317.1 | 3 - 4 | 1491 | 63 | |
NM_053167.3 | NP_444397.2 | 3 - 4 | 1491 | 63 | |
GenBank mRNA | AF220039.1 | - | 1 - 2 | 99 | 63 |
AK028280.1 | - | 3 - 4 | 1210 | 63 | |
AK035807.1 | - | 3 - 4 | 1209 | 63 | |
AK048875.1 | - | 3 - 4 | 1228 | 63 | |
AK049907.1 | - | 3 - 4 | 1205 | 63 | |
AK129109.1 | - | 3 - 4 | 1196 | 63 | |
AK163123.1 | - | 3 - 4 | 1228 | 63 | |
BC052034.1 | - | 3 - 4 | 1491 | 63 |