Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl21c
Entrez Gene ID
433294
Gene Name
methyltransferase like 21C
Gene Aliases
A530098C11Rik
Chromosome Location
Chr.1: 44009399 - 44042626 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013799.1 NP_001013821.1 3 - 4 343 149
GenBank mRNA AK041301.1 - 3 - 4 343 149
BC147328.1 - 3 - 4 281 149
BC147329.1 - 3 - 4 281 149

View Full Product Details