The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144558.4 | NP_653141.3 | 1 - 2 | 305 | 91 |
GenBank mRNA | AF411386.1 | - | 2 - 3 | 307 | 91 |
AK034329.1 | - | 1 - 2 | 267 | 91 | |
AK047404.1 | - | 1 - 2 | 307 | 91 | |
BC082584.1 | - | 1 - 2 | 111 | 91 |