Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tek
Entrez Gene ID
21687
Gene Name
endothelial-specific receptor tyrosine kinase
Gene Aliases
AA517024, Cd202b, Hyk, STK1, Tie-2, Tie2
Chromosome Location
Chr.4: 94739289 - 94874976 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290549.1 NP_001277478.1 21 - 22 3536 71
NM_001290551.1 NP_001277480.1 20 - 21 3386 71
NM_013690.3 NP_038718.2 21 - 22 3539 71
GenBank mRNA BC050824.1 - 20 - 21 3380 71
D13738.1 - 21 - 22 3544 71
S67051.1 - 22 - 23 3199 71
X67553.1 - 21 - 22 3307 71
X71426.1 - 22 - 23 3270 71

View Full Product Details