The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029837.2 | NP_001025008.2 | 4 - 5 | 647 | 76 |
NM_001164049.1 | NP_001157521.1 | 4 - 5 | 674 | 76 | |
NM_001164050.1 | NP_001157522.1 | 4 - 5 | 674 | 76 | |
NM_001164051.1 | NP_001157523.1 | 4 - 5 | 698 | 76 | |
NM_001164052.1 | NP_001157524.1 | 2 - 3 | 401 | 76 | |
NM_008840.3 | NP_032866.2 | 5 - 6 | 849 | 76 | |
GenBank mRNA | AK040867.1 | - | 4 - 5 | 609 | 76 |
AK149935.1 | - | 5 - 6 | 748 | 76 | |
AK150649.1 | - | 4 - 5 | 575 | 76 | |
AK153304.1 | - | 4 - 5 | 609 | 76 | |
AK155835.1 | - | 4 - 5 | 560 | 76 | |
AK171025.1 | - | 4 - 5 | 657 | 76 | |
AK172222.1 | - | 4 - 5 | 551 | 76 | |
BC035203.1 | - | 4 - 5 | 664 | 76 | |
CB248733.1 | - | 4 - 5 | 570 | 76 |