Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gpr89
- Entrez Gene ID
-
67549
- Gene Name
- G protein-coupled receptor 89
- Gene Aliases
-
4933412D19Rik, Gpr89a, SH120
- Chromosome Location
-
Chr.3: 96871066 - 96905343 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK034931.1 |
- |
- |
655 |
148 |
View Full Product Details