The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007661.3 | NP_031687.2 | 2 - 3 | 202 | 123 |
GenBank mRNA | AK008283.1 | - | 2 - 3 | 179 | 123 |
AK077668.1 | - | 2 - 3 | 203 | 123 | |
AK135402.1 | - | 2 - 3 | 209 | 123 | |
AK147133.1 | - | 2 - 3 | 180 | 123 | |
L37092.1 | - | 1 - 2 | 777 | 123 |