Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc4a3
Entrez Gene ID
20536
Gene Name
solute carrier family 4 (anion exchanger), member 3
Gene Aliases
A930038D23Rik, Ae3
Chromosome Location
Chr.1: 75546041 - 75559431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009208.3 NP_033234.2 19 - 20 3227 72
GenBank mRNA AK147218.1 - - 3564 72
BC066086.1 - 20 - 21 3101 72
BC080271.1 - 19 - 20 3222 72
M28383.1 - 19 - 20 3197 72

View Full Product Details