Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc4a3
Entrez Gene ID
20536
Gene Name
solute carrier family 4 (anion exchanger), member 3
Gene Aliases
A930038D23Rik, Ae3
Chromosome Location
Chr.1: 75546041 - 75559431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009208.3 NP_033234.2 5 - 6 829 95
GenBank mRNA AK045583.1 - 4 - 5 716 95
AK147218.1 - - 841 95
BC066086.1 - 6 - 7 688 95
BC080271.1 - 5 - 6 824 95
M28383.1 - 5 - 6 799 95

View Full Product Details