The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081456.1 | NP_001074925.1 | 8 - 9 | 1291 | 68 |
NM_148937.2 | NP_683739.2 | 8 - 9 | 1442 | 68 | |
GenBank mRNA | AK016945.1 | - | 8 - 9 | 1439 | 68 |
AK029561.1 | - | 8 - 9 | 1437 | 68 | |
AK039149.1 | - | 8 - 9 | 1269 | 68 | |
AK140231.1 | - | 8 - 9 | 1278 | 68 | |
AY033991.1 | - | 8 - 9 | 1274 | 68 |