The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153744.3 | NP_714966.1 | 13 - 14 | 1692 | 104 |
GenBank mRNA | AK036585.1 | - | 13 - 14 | 1693 | 104 |