The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146299.1 | NP_001139771.1 | 1 - 2 | 486 | 69 |
NM_172966.3 | NP_766554.2 | 1 - 2 | 486 | 69 | |
GenBank mRNA | AK033601.1 | - | 1 - 2 | 530 | 69 |
BC137955.1 | - | 1 - 2 | 439 | 69 | |
BC145060.1 | - | 1 - 2 | 439 | 69 |