Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptprh
Entrez Gene ID
545902
Gene Name
protein tyrosine phosphatase, receptor type, H
Gene Aliases
sap-1
Chromosome Location
Chr.7: 4548614 - 4604041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207270.2 NP_997153.2 11 - 12 2257 59
GenBank mRNA AB217856.1 - 10 - 11 1930 59
AK078884.1 - 10 - 11 1934 59

View Full Product Details