Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shoc2
Entrez Gene ID
56392
Gene Name
soc-2 (suppressor of clear) homolog (C. elegans)
Gene Aliases
AU017287, Sur8, mKIAA0862
Chromosome Location
Chr.19: 53944232 - 54033280 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168505.1 NP_001161977.1 5 - 6 1481 129
NM_019658.6 NP_062632.2 5 - 6 1522 129
GenBank mRNA AF068921.1 - 4 - 5 1157 129
AK146447.1 - 5 - 6 1519 129
BC026364.1 - 4 - 5 1388 129
BC049775.1 - 4 - 5 1387 129
BC083060.1 - 5 - 6 1481 129

View Full Product Details