Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc15a5
Entrez Gene ID
277898
Gene Name
solute carrier family 15, member 5
Gene Aliases
9830102E05Rik
Chromosome Location
Chr.6: 137983590 - 138079916 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177787.4 NP_808455.2 6 - 7 1323 72
GenBank mRNA AK036413.1 - 5 - 6 1106 72
BC026899.1 - 3 - 4 401 72
BC039761.1 - 3 - 4 412 72
BC141332.1 - 5 - 6 1042 72
BC141334.1 - 5 - 6 1042 72

View Full Product Details