Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Neto1
Entrez Gene ID
246317
Gene Name
neuropilin (NRP) and tolloid (TLL)-like 1
Gene Aliases
AI851453, Btcl1, C130005O10Rik
Chromosome Location
Chr.18: 86394952 - 86506721 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144946.4 NP_659195.3 8 - 9 1909 87
GenBank mRNA AF448840.1 - 8 - 9 1242 87
AK032510.1 - 6 - 7 777 87
AK043710.1 - 8 - 9 1245 87
AK081325.1 - 8 - 9 1910 87
AY138990.1 - 8 - 9 978 87
BC051145.1 - 8 - 9 1269 87

View Full Product Details