The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201353.1 | NP_958741.1 | 1 - 2 | 222 | 79 |
GenBank mRNA | AK141581.1 | - | 1 - 2 | 246 | 79 |
BC057070.1 | - | 1 - 2 | 222 | 79 |