Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nhej1
- Entrez Gene ID
-
75570
- Gene Name
- nonhomologous end-joining factor 1
- Gene Aliases
-
1700029B21Rik, XLF, cernunnos
- Chromosome Location
-
Chr.1: 74967346 - 75046653 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK006481.1 |
- |
- |
67 |
150 |
View Full Product Details