Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp2j13
Entrez Gene ID
230459
Gene Name
cytochrome P450, family 2, subfamily j, polypeptide 13
Gene Aliases
-
Chromosome Location
Chr.4: 96042660 - 96077566 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145548.4 NP_663523.2 7 - 8 1269 63
GenBank mRNA AK143903.1 - 7 - 8 1270 63
AK143981.1 - 7 - 8 1266 63
AK162358.1 - - 1273 63
AK165405.1 - 7 - 8 1269 63
AK165469.1 - 7 - 8 1247 63
AK165473.1 - 7 - 8 1247 63

View Full Product Details