Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spink10
Entrez Gene ID
328971
Gene Name
serine peptidase inhibitor, Kazal type 10
Gene Aliases
A230091H23Rik
Chromosome Location
Chr.18: 62548911 - 62662933 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177829.3 NP_808497.1 7 - 8 577 92
GenBank mRNA AK039062.1 - 7 - 8 578 92
AV380905.2 - 2 - 3 113 92
BC049733.1 - - 234 92
BC125360.1 - 7 - 8 493 92
BC125362.1 - 7 - 8 493 92
DQ437330.1 - 7 - 8 577 92

View Full Product Details