Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc68
Entrez Gene ID
381175
Gene Name
coiled-coil domain containing 68
Gene Aliases
BC046343
Chromosome Location
Chr.18: 69924858 - 69969484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201362.2 NP_958750.1 5 - 6 541 62
GenBank mRNA AK078964.1 - 5 - 6 543 62
BC046343.1 - 5 - 6 521 62

View Full Product Details