The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010773.2 | NP_034903.2 | 1 - 2 | 676 | 67 |
GenBank mRNA | AF072243.1 | - | 1 - 2 | 634 | 67 |
AF072245.1 | - | 1 - 2 | 634 | 67 | |
AK147008.1 | - | 1 - 2 | 561 | 67 | |
BC026698.1 | - | 1 - 2 | 636 | 67 | |
BC046607.1 | - | 1 - 2 | 676 | 67 | |
BC057289.1 | - | 1 - 2 | 508 | 67 | |
BC058864.1 | - | 1 - 2 | 496 | 67 |