The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010545.3 | NP_034675.1 | 6 - 7 | 659 | 109 |
GenBank mRNA | AK002232.1 | - | 6 - 7 | 659 | 109 |
AK020607.1 | - | 6 - 7 | 659 | 108 | |
BC096435.1 | - | 6 - 7 | 670 | 109 | |
X00496.1 | - | 5 - 6 | 436 | 109 |