The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042605.1 | NP_001036070.1 | 8 - 9 | 916 | 120 |
NM_010545.3 | NP_034675.1 | 7 - 8 | 724 | 120 | |
GenBank mRNA | AK002232.1 | - | 7 - 8 | 724 | 120 |
AK020607.1 | - | 7 - 8 | 724 | 120 | |
AK154096.1 | - | 8 - 9 | 947 | 120 | |
BC096435.1 | - | 7 - 8 | 735 | 120 | |
X00496.1 | - | 6 - 7 | 501 | 120 |