Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgef37
Entrez Gene ID
328967
Gene Name
Rho guanine nucleotide exchange factor (GEF) 37
Gene Aliases
4933429F08Rik
Chromosome Location
Chr.18: 61493625 - 61537044 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177828.4 NP_808496.2 2 - 3 342 101
GenBank mRNA AB199795.1 - 1 - 2 192 101
CJ045695.1 - 2 - 3 101
CJ131942.1 - 2 - 3 243 101

View Full Product Details