The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007859.4 | NP_031885.3 | 1 - 2 | 222 | 82 |
GenBank mRNA | AB009377.1 | - | 1 - 2 | 130 | 82 |
AK036094.1 | - | 1 - 2 | 199 | 82 | |
AK077619.1 | - | 1 - 2 | 268 | 82 | |
AK089267.1 | - | 1 - 2 | 199 | 82 | |
BC053052.1 | - | 1 - 2 | 152 | 82 |