Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfyve9
Entrez Gene ID
230597
Gene Name
zinc finger, FYVE domain containing 9
Gene Aliases
E030027L17, Madhip, NSP, SARA, SMADIP
Chromosome Location
Chr.4: 108637834 - 108781148 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183300.2 NP_899123.2 7 - 8 920 124
GenBank mRNA DQ303413.1 - 7 - 8 920 124

View Full Product Details