The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147221.2 | NP_671754.2 | 2 - 3 | 482 | 80 |
GenBank mRNA | AF486579.1 | - | 2 - 3 | 481 | 80 |