Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Orc1
Entrez Gene ID
18392
Gene Name
origin recognition complex, subunit 1
Gene Aliases
AA545195, MmORC1, Orc1l
Chromosome Location
Chr.4: 108575499 - 108616048 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011015.2 NP_035145.2 2 - 3 225 85
GenBank mRNA AB190255.1 - 1 - 2 92 85
AJ003133.1 - 2 - 3 215 85
AK042130.1 - 2 - 3 211 85
AK141741.1 - 2 - 3 228 85
AK145260.1 - 2 - 3 219 85
AK161814.1 - 2 - 3 226 85
AK164035.1 - 2 - 3 226 85
BC015073.1 - 2 - 3 151 85

View Full Product Details