Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otop3
Entrez Gene ID
69602
Gene Name
otopetrin 3
Gene Aliases
2310011E08Rik
Chromosome Location
Chr.11: 115334731 - 115346927 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027132.2 NP_081408.2 3 - 4 716 77
GenBank mRNA AB087505.1 - 3 - 4 671 77
AK043266.1 - 3 - 4 671 77
BC145613.1 - 3 - 4 633 77
BC150803.1 - 3 - 4 633 77

View Full Product Details