Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hif3a
Entrez Gene ID
53417
Gene Name
hypoxia inducible factor 3, alpha subunit
Gene Aliases
Ipas, MOP7, NEPAS, bHLHe17
Chromosome Location
Chr.7: 17030993 - 17062427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016868.3 NP_058564.2 1 - 2 103 69
GenBank mRNA AF060194.1 - 1 - 2 103 69
BC120587.1 - 1 - 2 56 69

View Full Product Details