Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pik3cg
Entrez Gene ID
30955
Gene Name
phosphoinositide-3-kinase, catalytic, gamma polypeptide
Gene Aliases
5830428L06Rik, PI3Kgamma, p110gamma
Chromosome Location
Chr.12: 32173129 - 32208677 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146200.1 NP_001139672.1 8 - 9 2904 89
NM_001146201.1 NP_001139673.1 8 - 9 2960 89
NM_020272.2 NP_064668.2 8 - 9 3081 89
GenBank mRNA AF208345.1 - 7 - 8 2760 89
AJ249280.1 - 8 - 9 3081 89
AK040543.1 - 8 - 9 2866 89
AK077772.1 - 2 - 3 236 89
AK132553.1 - 8 - 9 2906 89
AK137470.1 - 8 - 9 2813 89
AK137620.1 - 8 - 9 2962 89
BC051246.1 - 8 - 9 2821 89

View Full Product Details