Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gm9926
- Entrez Gene ID
-
100503565
- Gene Name
- predicted gene 9926
- Gene Aliases
-
ENSMUSG00000053891
- Chromosome Location
-
Chr.18: 66504178 - 66511737 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK033454.1 |
- |
1 - 2 |
284 |
109 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB624834.1 |
- |
1 - 2 |
281 |
|
Chr. 18 - 66504183 - 66507575 [+] on Build GRCm38 |
109 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
phorbol-12-myristate-13-acetate-induced protein 1 |
Pmaip1 |
Noxa |
-38.6kb
|
Sense |
Chr. - 66458604 - 66465558 on Build GRCm38 |
Downstream |
melanocortin 4 receptor |
Mc4r |
Mc4-r |
352.3kb
|
Anti-sense |
Chr. - 66857705 - 66860487 on Build GRCm38 |
View Full Product Details