Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casc1
Entrez Gene ID
320662
Gene Name
cancer susceptibility candidate 1
Gene Aliases
A230084G12Rik, Las1
Chromosome Location
Chr.6: 145174872 - 145210970 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177222.4 NP_796196.3 3 - 4 265 80
GenBank mRNA AK132912.1 - 3 - 4 489 80
AY423542.1 - 3 - 4 196 80
AY485607.1 - 3 - 4 265 80
AY485608.1 - 3 - 4 265 80
BC072568.1 - 1 - 2 107 80
BC120743.1 - 3 - 4 169 80
BC125423.1 - 3 - 4 169 80
BC144801.1 - 3 - 4 169 80

View Full Product Details