The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081223.2 | NP_001074692.1 | 17 - 18 | 2890 | 88 |
NM_001252495.1 | NP_001239424.1 | 17 - 18 | 2795 | 88 | |
NR_045526.1 | 16 - 17 | 2763 | 88 | ||
NR_045527.1 | 16 - 17 | 2734 | 88 | ||
GenBank mRNA | AK037049.1 | - | 17 - 18 | 2813 | 88 |
AK041384.1 | - | 16 - 17 | 2640 | 88 | |
AK077614.1 | - | 16 - 17 | 2842 | 88 | |
AK163908.1 | - | - | 2730 | 88 | |
BC019665.1 | - | 14 - 15 | 2235 | 88 | |
BC050849.1 | - | 17 - 18 | 2795 | 88 | |
CX567593.1 | - | 3 - 4 | 327 | 88 |