Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbbp8
Entrez Gene ID
225182
Gene Name
retinoblastoma binding protein 8
Gene Aliases
9930104E21Rik, CtIP, RBBP-8, RIM, SAE2
Chromosome Location
Chr.18: 11633276 - 11743207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081223.2 NP_001074692.1 12 - 13 2373 81
NM_001252495.1 NP_001239424.1 12 - 13 2278 81
NR_045527.1 11 - 12 2217 81
GenBank mRNA AK037049.1 - 12 - 13 2296 81
AK041384.1 - 11 - 12 2123 81
AK077614.1 - 11 - 12 2325 81
BC019665.1 - 9 - 10 1718 81
BC027367.1 - 12 - 13 2241 81
BC050849.1 - 12 - 13 2278 81

View Full Product Details