Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdzk1
Entrez Gene ID
59020
Gene Name
PDZ domain containing 1
Gene Aliases
1700023D20Rik, 2610507N21Rik, 4921513F16Rik, AI267131, AI314638, AL022680, D3Ertd537e, Pdzd1, mPDZK1
Chromosome Location
Chr.3: 96830065 - 96870926 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021517.2 NP_067492.2 1 - 2 88 132
GenBank mRNA AK012070.1 - 1 - 2 78 132
AW259368.1 - 1 - 2 88 132

View Full Product Details