Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog5
Entrez Gene ID
238123
Gene Name
component of oligomeric golgi complex 5
Gene Aliases
5430405C01Rik, C87247, D18362, GOLTC1, GTC90
Chromosome Location
Chr.12: 31654868 - 31937630 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163126.1 NP_001156598.1 1 - 2 114 122
GenBank mRNA AK030656.1 - 1 - 2 115 122

View Full Product Details