The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168491.1 | NP_001161963.1 | 11 - 12 | 1550 | 97 |
NM_001168492.1 | NP_001161964.1 | 11 - 12 | 1550 | 97 | |
NM_011050.4 | NP_035180.2 | 11 - 12 | 1638 | 97 | |
GenBank mRNA | AK134366.1 | - | 11 - 12 | 1639 | 97 |
AK172654.1 | - | 11 - 12 | 1506 | 97 | |
BC055739.1 | - | 11 - 12 | 1485 | 97 | |
CT010188.1 | - | 10 - 11 | 1348 | 97 | |
D50465.1 | - | 10 - 11 | 1418 | 97 | |
D86344.1 | - | 11 - 12 | 1434 | 97 |