Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dvl1
Entrez Gene ID
13542
Gene Name
dishevelled, dsh homolog 1 (Drosophila)
Gene Aliases
Dvl, mKIAA4029
Chromosome Location
Chr.4: 155847317 - 155859303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK155349.1 - 4 - 5 772 51
AK220168.1 - 3 - 4 960 51
BC051991.1 - 4 - 5 531 51
BC088987.1 - 4 - 5 636 51
BC138848.1 - 5 - 6 643 51
BC138849.1 - 5 - 6 643 51
BU702464.1 - 4 - 5 433 51
U10115.1 - 4 - 5 729 51

View Full Product Details