Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ttll10
- Entrez Gene ID
-
330010
- Gene Name
- tubulin tyrosine ligase-like family, member 10
- Gene Aliases
-
4833412E22Rik, 4930595O22Rik, AU022799, BC042775, Ttll5
- Chromosome Location
-
Chr.4: 156023748 - 156084440 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK014686.1 |
- |
1 - 2 |
349 |
106 |
View Full Product Details