The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011659.2 | NP_035789.1 | 5 - 6 | 808 | 82 |
GenBank mRNA | BC065782.1 | - | 5 - 6 | 733 | 82 |
BC139239.1 | - | 5 - 6 | 678 | 82 | |
BC139266.1 | - | 5 - 6 | 678 | 82 | |
Z21674.1 | - | 5 - 6 | 629 | 82 |