The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028020.3 | NP_082296.1 | 11 - 12 | 1257 | 72 |
GenBank mRNA | AK010425.1 | - | 11 - 12 | 1219 | 72 |
AK090206.1 | - | 11 - 12 | 1258 | 72 | |
AK150436.1 | - | 11 - 12 | 1254 | 72 | |
AK152740.1 | - | 11 - 12 | 1254 | 72 | |
AK167607.1 | - | 11 - 12 | 1260 | 72 | |
AK172533.1 | - | 11 - 12 | 1241 | 72 | |
BC008240.1 | - | 3 - 4 | 353 | 72 | |
BC011155.1 | - | 11 - 12 | 1241 | 72 |